Learn More
Invitrogen™ CellROX™ Reagent Variety Pack, for oxidative stress detection
For oxidative stress detection
Supplier: Invitrogen™ C10448
Description
The CellROX Variety Pack contains one vial (50μL) each of CellROXGreen Reagent, CellROXOrange Reagent, and CellROXDeep Red Reagent. These reagents are novel fluorogenic probes for measuring cellular oxidative stress in live cells. These cell-permeant dyes exhibit bright fluorescence upon oxidation by reactive oxygen species (ROS).
- Optimized for detection of oxidative stress in live cells and may be added directly to serum-containing growth media
- Provided as stable, ready-to-use DMSO solutions with simple protocols, compatible with standard workflows in fluorescence microscopy
- Compatible with a variety of detection platforms, including traditional fluorescence microscopy, high content screening (HCS), and flow cytometry
- Oxidative stress results from an imbalance between production of reactive oxygen species (ROS) and ability of cells to scavenge them
- ROS play an important role in progression of several diseases, including inflammation, atherosclerosis, aging, and age-related degenerative disorders
- Form: Frozen
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Wet Ice
Specifications
Orange, Far-Red, Green | |
Frozen | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.