missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Click-iT™ Protein Reaction Buffer Kit DFS Item

Click two moieties together: azide or alkyne and the corresponding click detection reagent

Supplier:  Invitrogen™ C10276

Catalog No. C10276


Explore available promotions

Description

Description

The Click-iT Protein Buffer Kit provides researchers who have proteins labeled with an azide or alkyne and the corresponding click detection reagent with the means to click these two moieties together. Each kit includes sufficient material to perform 25 reactions for subsequent analysis by gel electrophoresis, western blot or mass spectrometry.

  • Less hazardous

Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Less hazardous
25 Reactions kit
Room Temperature
Protein Reaction Buffer Kit
Store at room temperature.
Click-iT™, Molecular Probes™
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.