missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DAF-FM (4-Amino-5-Methylamino-2',7'-Difluorofluorescein) DFS Item

Use to detect and quantify low concentrations of nitric oxide

Supplier:  Invitrogen™ D23841

Catalog No. D23841


Explore available promotions

Description

Description

DAF-FM is essentially nonfluorescent until it reacts with NO to form fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

  • Ex/Em of DAF-FM: approx. 495/515nm
  • Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
  • DAF-FM can be introduced into cells by pressure injection or perfusion from patch-clamp pipette
  • Buffers containing bovine serum albumin (BSA) and phenol red may affect the fluorescence and should be used with caution
  • Fluorescence quantum yield of DAF-FM is ∽0.005, but increases about 160-fold, to ∽0.81, after reacting with NO
  • Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
  • Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
  • in vivo imaging of NO in zebrafish
  • Intravital microscopic detection of NO generation associated with angiogenesis in mice
  • Quantitation of ATP-induced NO release in rabbit platelets
  • Spectra of NO adduct of DAF-FM are independent of pH above pH 5.5
  • NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture
  • DAF-FM is more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM approx. 3nM versus approx. 5nM for DAF-2)

Refer to the Molecular Probes™ Handbook for additional product information.

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

1 mg
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.