missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DDAO Phosphate (9H-(1,3-Dichloro-9,9-Dimethylacridin-2-One-7-yl) Phosphate, Diammonium Salt) DFS Item

Easily distinguish substrate and hydrolysis product

Supplier:  Invitrogen™ D6487

Catalog No. D6487


Explore available promotions

Description

Description

The phosphatase substrate DDAO phosphate [9H-(1,3-dichloro-9,9-dimethylacridin-2-one-7-yl) phosphate, diammonium salt] yields a hydrolysis product that can be excited with the 633nm laser (excitation/emission maxima approx. 645/660). Although the substrate itself is fluorescent (excitation/emission maxima approx. 460/610nm), the difference between the substrate excitation maximum and that of its hydrolysis product is over 200nm, allowing the two species to be easily distinguished.

Cell Analysis, Drug Discovery and Development, Enzyme and Protein Activity Assays, Labeling Chemistry, Labeling QC and Calibration, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Powder
Phosphatase Substrate
Phosphatases
Fluorometer, Microplate Reader
Room Temperature
Fluorescence
5 mg
Chemical Substrate
646⁄659
DDAO
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.