missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DQ™ Ovalbumin - Special Packaging DFS Item

Suited for antigen processing and presentation studies

Supplier:  Invitrogen™ D12053

Catalog No. D12053


Explore available promotions

Description

Description

DQ ovalbumin is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ ovalbumin to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.

Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Solid
5 x 1 mg
Protein-Based Substrate
BODIPY™ FL
Room Temperature
Fluorescence
DQ™
Protease Substrate
Proteases
Antigen processing and presentation studies
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.