missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ DQ™ Red BSA - Special Packaging
Hydrolysis of substrate yields brightly fluorescent product
Supplier: Invitrogen™ D12051
Description
DQ Red BSA is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ Red BSA to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.
Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Solid | |
5 x 1 mg | |
Protein-Based Substrate |
Fluorescence | |
DQ™ | |
Protease Substrate | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.