missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ EnzChek™ Lipase Substrate, green fluorescent, 505/515 DFS Item

Green-fluorescent product

Supplier:  Invitrogen™ E33955

Catalog No. E33955


Explore available promotions

Description

Description

The nonfluorescent EnzChek™ lipase substrate, in the presence of lipases, produces a green-fluorescent product (excitation/emission maxima ∼507/515) for accurate lipase detection in solution.

Dissolve in high-quality anhydrous DMSO or chloroform to prepare a stock concentrations of up to 1 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store in freezer (-5°C to -30°C) and protect from light.
Solid
100 μg
Chemical Substrate
1011.15 g/mol
Fluorescence
EnzChek™
Lipase substrate
507/515 nm
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.