missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ FluoCells™ Prepared Slide #1 (BPAE cells with MitoTracker™ Red CMXRos, Alexa Fluor™ 488 Phalloidin, and DAPI) DFS Item

Contains fixed, multi-labeled, cell preparations for observation by fluorescence microscopy validated for immunofluorescence

Supplier:  Invitrogen™ F36924

Catalog No. F36924

Explore available promotions

Description

Description

FluoCells Prepared Slide #1 contains bovine pulmonary artery endothelial cells (BPAEC). MitoTracker Red CMXRos was used to stain the mitochondria in the live cells, with accumulation dependent upon membrane potential. Following fixation and permeabilization, F-actin was stained with Alexa Fluor 488 phalloidin, and the nuclei were counterstained with the blue-fluorescent DNA stain DAPI.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Prepared Microscope Slide
Immunofluorescence
1 Ea.
Alexa Fluor FluoCells MitoTracker
Store at room temperature and protect from light.
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.