missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ FluoZin™-2, AM, cell permeant
Designed to detect Zn2+ concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists
Supplier: Invitrogen™ F24189
Description
- Based on N-(2-methoxyphenyl)iminodiacetate chelator,designed for detection of Zn2+ in the 0-100μM range with minimal interfering Ca2+ sensitivity
- Label (Ex/Em): FluoZin-2 (approximately 495/525nm)
- Kd (Zn2+) (in buffer): approximately 2μM
- Lyophilized product may be dissolved in DMSO for use
- Product is loaded into cells by adding the dissolved indicator to medium containing cells
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, Zinc and Heavy Metal Ion Detection
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Zinc Indicator | |
Room Temperature |
Cell Viability and Proliferation | |
FluoZin™ |
For Research Use Only. Not for use in diagnostic procedures.