Invitrogen™ Griess Reagent Kit, for nitrite quantitation DFS Item
For nitrite quantitation
Supplier:  Invitrogen™ G7921
Catalog No. G7921
Explore available promotions
Description
Description
  • Greiss reagent kit provides simple and well characterized colorimetric assay for nitrites, and nitrates that have been reduced to nitrites, with detection limit of about 100nM
Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Nitric Oxide Research, Nitro-Oxidative Stress
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
1 kit
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.