missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ JC-9 Dye (Mitochondrial Membrane Potential Probe) DFS Item

Mitochondrial membrane potential probe

Supplier:  Invitrogen™ D22421

Catalog No. D22421


Explore available promotions

Description

Description

The ratiometric mitochondrial membrane potential probe JC-9 exhibits green fluorescence that is independent of membrane potential and an increase in red fluorescence at hyperpolarized membrane potentials.

Apoptosis, Cell Analysis, Cell Structure, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Mitochondria, Mitochondria Function, Mitochondrial Function

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Green
Fluorescence
Mitochondrial Membrane Potential Probe
Room Temperature
Store at room temperature and protect from light.
Fluorescence Microscope, Flow Cytometer
5 mg
Mitochondria
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.