Learn More
Invitrogen™ LIVE/DEAD™ Yeast Viability Kit
Viability required to change green intracelluar stain into red-orange intravacuolar structures
Supplier: Invitrogen™ L7009
Description
The LIVE/DEAD Yeast Viability Kit combines a novel two-color fluorescent probe for yeast viability, FUN™ 1, with a fluorescent fungal surface labeling reagent, Calcofluor™ White M2R. Plasma membrane integrity and metabolic function of fungi are required to convert the yellow-green-fluorescent intracellular staining of FUN 1 into red-orange intravacuolar structures; Calcofluor White M2R labels cell-wall chitin with blue-fluorescence regardless of metabolic state.
Analysis of Yeast, Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Microbiological Analysis, Yeast Viability and Cytotoxicity
Order Info
Shipping Condition: Room temperature
Specifications
Yeast Cells | |
Store in freezer at -5°C to -30°C and protect from light. | |
Fluorescence | |
Fluorescence Microscope, Fluorometer, Flow Cytometer, Microplate Reader | |
Other Label(s) or Dye(s) | |
485, 450 nm | |
LIVE/DEAD™ | |
Room Temperature |
Yellow-green, red-orange, blue | |
LIVE/DEAD™ Yeast Viability Kit | |
Viability Assay | |
Yeast Viability Kit | |
530, 620, 500, 550 nm | |
Tube(s), 96-well plate, Slide(s) | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.