Invitrogen™ NanoOrange™ Protein Quantitation Kit, 200-2,000 assays DFS Item
Sensitive and easy assay for protein quantitation, with detection as low as 10 ng/mL of protein in solution
Supplier:  Invitrogen™ N6666
Catalog No. N6666
Explore available promotions
Description
Description
  • Fluorescent dye is suitable for use with spectrofluorometers and microplate readers
  • For detection of lipoproteins or proteins in a complex lipid environment,use CBQCA Protein Quantitation Kit (C-6667)
Protein Quantitation, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
Fluorescent Protein Assay
NanoOrange™
200 to 2000 Assays
Microplate Reader
Fluorescence
Protein Quantitation Assay
Protein quantitation
1 Kit
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.