missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Nitro blue Tetrazolium Chloride (NBT) DFS Item

Use to assess cell viability as function of redox potential

Supplier:  Invitrogen™ N6495

Catalog No. N6495


Explore available promotions

Description

Description

  • Actively respiring cells convert water-soluble Nitro blue tetrazolium chloride to a deep blue-colored precipitate

Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunohistochemistry (IHC), Nitro-Oxidative Stress, Proteins, Expression, Isolation and Analysis, Reactive Oxygen Species, Western Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature and protect from light.
1 g
Chemical Substrate
Colorimetric
Oxidase Substrate
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.