missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospholipase C Protein, Phosphatidylinositol-Specific (from Bacillus cereus)
Description
This phosphatidylinositol-specific phospholipase C is a highly purified enzyme preparation from Bacillus cereus which cleaves phospholipid phosphatidylinositol (PI) into two molecules. Validated for assay standard. Alphabetical protein filter: P
Cell Analysis, Cell Structure, Enzyme & Protein Activity Assays, Lipase & Phopholipase Activity, Membranes (General) & Lipids, Plasma Membrane, Proteins, Expression, Isolation & Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Concentration | 100 U⁄ml |
| For Use With (Application) | Assay Standard |
| Quantity | 50 μL |
| Source | Bacillus cereus |
| Species | B. cereus |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Shipping Condition | Wet Ice |
| Form | Solution |
| Protein Subtype | Other Proteins |
| Research Category | Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.