missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Quant-iT™ Protein Assay Kit
For easy and accurate protein quantitation
Supplier: Invitrogen™ Q33210
Description
- Kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards
- Simply dilute reagent, load into wells of microplate, add 1-20μL of sample, mix, and measure fluorescence
- Assay is highly selective for protein and exhibits little protein-to-protein variation
- Assay is performed at room temperature, and signal is stable for 3hours
- Common contaminants, such as salts, solvents, or DNA—but not detergents—are well tolerated in the assay
- Quant-iT DNA Assay Kits (Q33120, Q33130) and a Quant-iT RNA Assay Kit (Q33140) are also available
Protein Quantitation, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Fluorescence | |
Protein Quantitation Assay | |
3919448 | |
1 Kit | |
Room Temperature |
Fluorescent Protein Assay | |
Quant-iT™ | |
Microplate Reader | |
Concentrated assay reagent, dilution buffer, and pre-diluted BSA standards | |
1 to 20 uL | |
Store in refrigerator (2° to 8°C). Protect from light. |
For Research Use Only. Not for use in diagnostic procedures.