Invitrogen™ Singlet Oxygen Sensor Green - Special Packaging DFS Item
Detection reagent highly selective for singlet oxygen
Supplier:  Invitrogen™ S36002
Catalog No. S36002
Explore available promotions
Description
Description
Unlike other available fluorescent and chemiluminescent singlet oxygen detection reagents, Singlet Oxygen Sensor Green does not show any appreciable response to hydroxyl radical or superoxide.
  • Indicator initially exhibits weak blue fluorescence, but in presence of singlet oxygen, it emits green fluorescence (excitation/emission maxima approx. 504/525nm) similar to that of fluorescein
  • Singlet Oxygen Sensor Green is cell-impermeant
Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
10 x 100 μg
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.