missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ TetraSpeck™ Microspheres, 1.0 μm, fluorescent blue/green/orange/dark red DFS Item

Beads that each display four well-separated excitation/emission peaks

Supplier:  Invitrogen™ T7282

Catalog No. T7282


Explore available promotions

Description

Description

  • Stained throughout with four different fluorescent dyes
  • Excitation/emission peaks: 360/430nm (blue), 505/515nm (green), 560/580nm (orange) and 660/680nm (dark red)
  • Facilitate the calibration of conventional fluorescence microscopes, confocal laser scanning microscopes, and associated image-processing equipment for both scientific and commercial imaging, especially for multicolor applications

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Confocal Microscope Calibration, Fluorescence Microscope Calibration
Orange, Dark Red, Blue, Green
Suspension Beads
0.5 mL
1 μm
Store in refrigerator 2°C to 8°C and protect from light.
Microsphere
TetraSpeck
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.