missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum powder, -250 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

$72.14 - $72.14

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA0093206
View Documents
Thermo Scientific Chemicals
00093206
5 g
Each for $72.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]

Specifications

Gray
Mo
UN3089
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685
−250 Mesh
Powder
(metals basis)
7439-98-7
MFCD00003465
5 g
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
99.9%
Odorless
Molybdenum powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c

H228-H360-H371

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-107-2

RTECSNumber : QA4680000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only