Learn More
Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals
$81.19 - $374.48
Chemical Identifiers
CAS | 10241-05-1 |
---|---|
Molecular Formula | Cl5Mo |
Molecular Weight (g/mol) | 273.20 |
MDL Number | MFCD00003467 |
InChI Key | GICWIDZXWJGTCI-UHFFFAOYSA-I |
Synonym | molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h |
PubChem CID | 61497 |
ChEBI | CHEBI:30635 |
SMILES | [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC197080100
|
Thermo Scientific Chemicals
197080100 |
10 g | Glass Bottle |
Each for $81.19
|
|
||||
AC197081000
|
Thermo Scientific Chemicals
197081000 |
100 g | Glass bottle |
Each for $374.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
10241-05-1 | |
273.20 | |
GICWIDZXWJGTCI-UHFFFAOYSA-I | |
61497 | |
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5] |
Cl5Mo | |
MFCD00003467 | |
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h | |
CHEBI:30635 |
Specifications
268°C | |
10241-05-1 | |
94.0 | |
Fine Crystals or Crystalline Powder | |
94% min. (Argentometry) | |
MoCl5 | |
2508 | |
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h | |
GICWIDZXWJGTCI-UHFFFAOYSA-I | |
pentachloromolybdenum | |
61497 | |
273.2 | |
Glass Bottle |
194°C | |
Black to Blue or Black-Green | |
100.0 | |
10 g | |
Cl5Mo | |
MFCD00003467 | |
06,412; 01,1015; 02,365; 03,250; 04,428; 05,587; 06,519; 08,443; 09,414; 11,471; 12,434; 13,273; 04,99; 16,95 | |
Solubility in water: reacts with water. Other solubilities: soluble in dry ether,dry alcohol and other,organic solvents | |
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5] | |
273.20 | |
CHEBI:30635 | |
95% | |
Molybdenum(V) chloride, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 233-575-3
RTECSNumber : QA4690000
TSCA : TSCA
RUO – Research Use Only