Learn More
Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W), Thermo Scientific Chemicals
$207.99 - $2157.73
Chemical Identifiers
CAS | 1313-27-5 |
---|---|
Molecular Formula | MoO3 |
Molecular Weight (g/mol) | 143.95 |
MDL Number | MFCD00003469 |
InChI Key | JKQOBWVOAYFWKG-UHFFFAOYSA-N |
Synonym | molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide |
PubChem CID | 14802 |
ChEBI | CHEBI:30627 |
IUPAC Name | trioxomolybdenum |
SMILES | O=[Mo](=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1293014
|
Thermo Scientific Chemicals
01293014 |
25 g |
Each for $207.99
|
|
|||||
AA1293022
|
Thermo Scientific Chemicals
01293022 |
100 g |
Each for $589.64
|
|
|||||
AA1293036
|
Thermo Scientific Chemicals
01293036 |
500 g |
Each for $2,157.73
|
|
|||||
Description
It is used as a selective oxidation catalyst, and as a molybdenum oxidase enzyme. It is a source of molybdenum compounds and molybdenum metal which serves as an additive to steel and corrosion-resistant alloys. It is used for lubricants and lubricant additives. It has applications in process regulators, pigments, corrosion inhibitors, electrochemical devices, displays, and smoke suppressants. It finds use in analytical chemistry, ceramic glazes, enamels, energy storage, and in the manufacture of acrylonitrile through catalytic oxidation of ammonia and propene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-27-5 | |
143.95 | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
14802 | |
trioxomolybdenum |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
CHEBI:30627 | |
O=[Mo](=O)=O |
Specifications
1155°C (sublimation) | |
1313-27-5 | |
Powder | |
99.9995% (metals basis excluding W) | |
MFCD00003469 | |
Slightly soluble in water. Soluble in concentrated mineral acids,alkali hydroxide solutions. | |
O=[Mo](=O)=O | |
143.95 | |
CHEBI:30627 | |
Odorless | |
4.692 g/cm3 |
795°C | |
White to Gray | |
25 g | |
MoO3 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
trioxomolybdenum | |
14802 | |
143.94 | |
White to gray powder | |
Molybdenum(VI) oxide |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H335-H351
DOTInformation : Hazard Class: 6.1, Packing Group: III
EINECSNumber : 215-204-7
RTECSNumber : QA4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only