missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molybdenum wire, gold plated, 0.025mm (0.001in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Microelectronics, high temperature
Supplier: Thermo Scientific Chemicals 040671G6
Description
Microelectronics, high temperature
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Molybdenum wire, gold plated | |
7439-98-7 | |
Mo | |
10 m | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
99.95% | |
Wire |
0.025mm (0.001 in.) dia. | |
(metals basis) | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
≈0.005g/m (Mo only), 3 to 5 wt% Au plating |
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
CHEBI:28685 | |
[Mo] |
Safety and Handling
EINECSNumber : 231-107-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only