Learn More
Molybdenyl acetylacetonate, 97%, Thermo Scientific Chemicals
$202.05 - $839.34
Chemical Identifiers
CAS | 17524-05-9 |
---|---|
Molecular Formula | C10H14MoO6 |
Molecular Weight (g/mol) | 326.17 |
MDL Number | MFCD00011506 |
InChI Key | SKMUJBBRXZPAJY-VGKOASNMSA-L |
Synonym | 2, 4-Pentanedione, metal derivative |
IUPAC Name | dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) |
SMILES | O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC211140500
|
Thermo Scientific Chemicals
211140500 |
50 g | Glass bottle |
Each for $202.05
|
|
|||||
AC211142500
|
Thermo Scientific Chemicals
211142500 |
250 g | Glass bottle |
Each for $839.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
17524-05-9 | |
326.17 | |
SKMUJBBRXZPAJY-VGKOASNMSA-L | |
dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) |
C10H14MoO6 | |
MFCD00011506 | |
2, 4-Pentanedione, metal derivative | |
O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
184.0°C | |
Crystalline Powder | |
97% | |
MFCD00011506 | |
Solubility in water: 85g/L (20°C). Other solubilities: 0.31 g/100g alcohol (20°C), 1.84 g/100g benzene (20°C) | |
O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
326.17 | |
97% | |
Glass bottle |
17524-05-9 | |
50 g | |
C10H14MoO6 | |
2, 4-Pentanedione, metal derivative | |
SKMUJBBRXZPAJY-VGKOASNMSA-L | |
dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) | |
326.15 | |
Authentic | |
Molybdenyl acetylacetonate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 241-522-
RUO – Research Use Only