missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1053722

 View more versions of this product

Catalog No. AAA1053722


Explore available promotions

Description

Description

Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

Solubility
Soluble in dilute ammonium hydroxide, alkali and ammonia. Slightly soluble in water.

Notes
Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

Molybdic acid
100 g
MFCD00036287
Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water.
O[Mo](O)(=O)=O
161.96
CHEBI:25371
3.1
7782-91-4
H2MoO4
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
VLAPMBHFAWRUQP-UHFFFAOYSA-L
dihydroxy(dioxo)molybdenum
82208
MoO3 85% min

Chemical Identifiers

7782-91-4
161.96
VLAPMBHFAWRUQP-UHFFFAOYSA-L
82208
dihydroxy(dioxo)molybdenum
H2MoO4
MFCD00036287
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
CHEBI:25371
O[Mo](O)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c

H319-H335-H373

EINECSNumber : 231-970-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only