missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Monkey MRP2 Vesicles
Monkey MRP2 Vesicles are insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2). Our Monkey MRP2 Vesicles can be used to evaluate test compounds and drug interactions with MRP2 in in vitro assays.
Supplier: Invitrogen™ GM0018
Specifications
Frozen | |
Research Grade | |
Store at -80°C. | |
Dry Ice |
0.5 mL | |
Monkey | |
Vial |
For Research Use Only. Not for use in diagnostic procedures.