missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mono-benzyl malonate, 95%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 439800100

Catalog No. AC439800100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Mono-benzyl malonate
94.0
45°C to 48°C
Authentic
Glass bottle
MFCD00138132
mono-benzyl malonate, monobenzyl malonate, 3-benzyloxy-3-oxopropanoic acid, benzyl hemimalonate, benzyl hydrogen malonate, monobenzylmalonate, 2-benzyloxy carbonyl acetic acid, malonic acid monobenzyl ester, 3-benzyl-oxy-3-oxopropanoic acid, propanedioic acid, 1-phenylmethyl ester
OC(=O)CC(=O)OCC1=CC=CC=C1
194.19
CHEBI:84093
95%
40204-26-0
100.0
White
94% min. (HPLC)
C10H10O4
10 g
CFLAHQSWDKNWPW-UHFFFAOYSA-N
3-oxo-3-phenylmethoxypropanoic acid
181659
194.19
Crystalline Powder or Powder

Chemical Identifiers

40204-26-0
194.19
CFLAHQSWDKNWPW-UHFFFAOYSA-N
181659
3-oxo-3-phenylmethoxypropanoic acid
C10H10O4
MFCD00138132
mono-benzyl malonate, monobenzyl malonate, 3-benzyloxy-3-oxopropanoic acid, benzyl hemimalonate, benzyl hydrogen malonate, monobenzylmalonate, 2-benzyloxy carbonyl acetic acid, malonic acid monobenzyl ester, 3-benzyl-oxy-3-oxopropanoic acid, propanedioic acid, 1-phenylmethyl ester
CHEBI:84093
OC(=O)CC(=O)OCC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only