missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Monocaprylin | |
41°C | |
C11H22O4 | |
5 g | |
GHBFNMLVSPCDGN-UHFFFAOYSA-N | |
2,3-dihydroxypropyl octanoate | |
3033877 | |
218.29 | |
Crystalline Lumps |
26402-26-6 | |
White-Yellow | |
MFCD00056652 | |
Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol | |
CCCCCCCC(=O)OCC(CO)O | |
218.293 | |
CHEBI:85241 | |
≥98.0% (GC) |
Chemical Identifiers
26402-26-6 | |
218.293 | |
GHBFNMLVSPCDGN-UHFFFAOYSA-N | |
3033877 | |
2,3-dihydroxypropyl octanoate |
C11H22O4 | |
MFCD00056652 | |
Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol | |
CHEBI:85241 | |
CCCCCCCC(=O)OCC(CO)O |
Safety and Handling
EINECSNumber : (2)-0669
TSCA : Yes