Covidien Monoject™ Sharps Containers
Can be autoclaved or incinerated
$783.82 - $783.82
Catalog Number | Mfr. No. | Capacity (English) | Height (English) | Length (English) | Size | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Height (English) | Length (English) | Size | Price | Quantity | ||
22-652-092
|
Covidien
8881676236 |
4 qt. | 7 in. | 10.5 in. | Small |
N/A
|
|
||
Description
- Polished viewing windows on four sides
- Translucent colors with lead-free pigments containing no heavy metals
- Clear lid with extra-wide butterfly closure
- Features needle removal notches
- Red
Specifications
Red | |
6.75 in. |