Covidien Monoject™ Tube Caps
Easy-on, easy-off caps for glass blood collection tubes
Supplier:  Covidien Z1270019502
 View more versions of this product
Catalog No. 02-544-26
Explore available promotions
Description
Description
  • Exclusive thumb tab makes cap removal simple, quick and fatigue-free
  • Supplied in convenient, resealable and polybag-lined dispenser box
  • Ideal for benchtop use
  • Available in six colors to match the original blood tube stopper
  • Size: 13mm
Specifications
Specifications
Blue
13 mm glass blood collection tubes