missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Monolaurin | |
63°C | |
186°C | |
MFCD00037815 | |
monolaurin, 1-monolaurin, lauricidin, glyceryl laurate, glyceryl monolaurate, 1-glyceryl laurate, glycerol 1-laurate, 1-monolauroyl-rac-glycerol, 1-monododecanoylglycerol, glycerol monolaurate | |
CCCCCCCCCCCC(=O)OCC(CO)O | |
274.401 | |
CHEBI:75539 | |
≥98.0% (GC) |
142-18-7 | |
White | |
C15H30O4 | |
25 g | |
ARIWANIATODDMH-UHFFFAOYSA-N | |
2,3-dihydroxypropyl dodecanoate | |
14871 | |
274.40 | |
Crystalline Powder |
Chemical Identifiers
142-18-7 | |
274.401 | |
ARIWANIATODDMH-UHFFFAOYSA-N | |
14871 | |
2,3-dihydroxypropyl dodecanoate |
C15H30O4 | |
MFCD00037815 | |
monolaurin, 1-monolaurin, lauricidin, glyceryl laurate, glyceryl monolaurate, 1-glyceryl laurate, glycerol 1-laurate, 1-monolauroyl-rac-glycerol, 1-monododecanoylglycerol, glycerol monolaurate | |
CHEBI:75539 | |
CCCCCCCCCCCC(=O)OCC(CO)O |
Safety and Handling
EINECSNumber : (2)-0669
RTECSNumber : OF1600000
TSCA : Yes