missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Montelukast sodium, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 456440010

Catalog No. AC456440010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Montelukast sodium
1 g
MFCD00931431
LBFBRXGCXUHRJY-HKHDRNBDSA-M
sodium;2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate
23663996
608.17
115.0°C
151767-02-1
C35H35ClNNaO3S
Solubility in water: very soluble.
[Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
608.17
CHEBI:6993
98%
Soluble in water

Chemical Identifiers

151767-02-1
608.17
LBFBRXGCXUHRJY-HKHDRNBDSA-M
CHEBI:6993
[Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
C35H35ClNNaO3S
MFCD00931431
23663996
sodium;2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye damage.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only