missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Montmorillonite K10, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals L1516030

Catalog No. AAL1516030


Explore available promotions

Description

Description

Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

Solubility
Specifications

Specifications

Montmorillonite K10
250 g
MFCD00132796
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
GUJOJGAPFQRJSV-UHFFFAOYSA-N
dialuminum;dioxosilane;oxygen(2-);hydrate
71586775
1318-93-0
Al2H2O12Si4
14,6260
Insoluble in water.
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
360.307
Odorless

Chemical Identifiers

1318-93-0
360.307
GUJOJGAPFQRJSV-UHFFFAOYSA-N
71586775
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
Al2H2O12Si4
MFCD00132796
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
dialuminum;dioxosilane;oxygen(2-);hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-288-5

RTECSNumber : CT9450000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only