missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MOPS, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210237083

 View more versions of this product

Catalog No. ICN10237083


Explore available promotions

Description

Description

Specifications

Specifications

MOPS
White
250 g
C7H15NO4S
3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
1132-61-2
Powder
100.4%
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.3

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
CHEBI:44115
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-478-5

SDS
Documents

Documents

Promotions

Promotions