Learn More
Morpholin-3-one, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 460530050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Morpholin-3-one | |
Authentic | |
C4H7NO2 | |
5g | |
VSEAAEQOQBMPQF-UHFFFAOYSA-N | |
morpholin-3-one | |
66953 | |
98% |
109-11-5 | |
97.5% min. (GC) | |
MFCD00631009 | |
3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone | |
O=C1COCCN1 | |
101.11 | |
101.11 |
Chemical Identifiers
109-11-5 | |
101.11 | |
VSEAAEQOQBMPQF-UHFFFAOYSA-N | |
66953 | |
O=C1COCCN1 |
C4H7NO2 | |
MFCD00631009 | |
3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone | |
morpholin-3-one |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye damage. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
EINECSNumber : 203-647-9
RUO â Research Use Only