missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morpholine, 99+%, extra pure, Thermo Scientific Chemicals

Morpholine, 99+%, C4H9NO, CAS Number-110-91-8, 1-oxa-4-azacyclohexane, diethyleneimide oxide, diethylene imidoxide, diethylene oximide, diethylenimide oxide, drewamine, p-isoxazine, tetrahydro, tetrahydro-1,4-oxazine, tetrahydro-2h-1,4-oxazine, tetrahydro-p-oxazine, 1L, 99% min. (GC), 27, 5

Supplier:  Thermo Scientific Chemicals 158680025

 View more versions of this product

Catalog No. AC158680025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Morpholine
99.0
-5.0°C
126.0°C to 130.0°C
Authentic
Glass Bottle
1.4530 to 1.4550
2.5 L
01,705; 11,352; 13,14
15, 6362
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
C1COCCN1
87.12
2.23 mPa.s (20°C)
87.12
Extra Pure
110-91-8
100.0
0.9900g/mL
32°C
99% min. (GC)
C4H9NO
MFCD00005972
27, 5
0.99
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
YNAVUWVOSKDBBP-UHFFFAOYSA-N
morpholine
8083
CHEBI:34856
99+%
Liquid

Chemical Identifiers

110-91-8
87.12
YNAVUWVOSKDBBP-UHFFFAOYSA-N
8083
morpholine
C4H9NO
MFCD00005972
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
CHEBI:34856
C1COCCN1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

GHS Signal Word: Danger

EINECSNumber : 203-815-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only