missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morpholine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A103550E

 View more versions of this product

Catalog No. AAA103550E


Explore available promotions

Description

Description

Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.

Solubility
Miscible with water.      

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Morpholine
-5°C
129°C
99%
C4H9NO
MFCD00005972
UN2054
Hygroscopic
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
YNAVUWVOSKDBBP-UHFFFAOYSA-N
morpholine
8083
87.12
110-91-8
1.007
35°C (95°F)
Amine-like
1.454
2500 g
102549
14,6277
Miscible with water.
C1COCCN1
87.12
CHEBI:34856
99%

Chemical Identifiers

110-91-8
87.12
YNAVUWVOSKDBBP-UHFFFAOYSA-N
8083
morpholine
C4H9NO
MFCD00005972
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
CHEBI:34856
C1COCCN1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H311+H331-H314-H335-H373

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: MORPHOLINE

EINECSNumber : 203-815-1

RTECSNumber : QD6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only