missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ IL-17A (homodimer) Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IL-17AA homodimer protein levels from samples including serum, and cell culture supernatants.

$410.00 - $1450.00

Specifications

Accession Number Q62386
Assay Range 4-500 pg/mL
Assay Sensitivity 4 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Alert:

Contains Azide


Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125203
View Documents
Invitrogen™
88737176
20 x 96 Tests
Each for $1,450.00
 
501128984
View Documents
Invitrogen™
88737186
10 x 96 Tests
Each for $1,044.00
 
501128965
View Documents
Invitrogen™
88737122
2 x 96 Tests
Each for $410.00
 
Description

Description

The Mouse Interleukin 17A Homodimer (IL-17A) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17A. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Mouse IL-17A ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)
Specifications

Specifications

Q62386
4 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
Il17a
HRP
Serum, 100 μL; Supernatant, 100 μL
Interleukin-17A, IL-17AA
1 hr. 45 min.
4-500 pg/mL
Biotin
Serum, Supernatant
ELISA
16171
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only