Kent Scientific Mouse Surgical Kit DFS Item
Includes everything required to carry out surgery on a mouse.
Supplier:  Kent Scientific INSMOUSEKIT
Catalog No. 13-005-204
Explore available promotions
Description
Description
  • Includes: 11cm long Stainless steel No.5 Tweezer, 12.5cm long straight serrated needle holder, 8cm long straight McPherson-Vannas Scissors, Wire Speculum, 10cm long straight Dissecting Scissors, 10cm long serrated Iris Forceps, 14cm long Stainless Steel Blunt probe (dia.: 1mm), leather storage case
Specifications
Specifications
Mouse
Mouse Surgical Kit
11cm Stainless steel No.5 Tweezer, 12.5cm straight serrated needle holder, 8cm straight McPherson-Vannas Scissors, Wire Speculum, 10cm straight Dissecting Scissors, 10cm serrated Iris Forceps, 14cm Stainless Steel Blunt probe