Learn More
Kent Scientific MouseSTAT™ Rat Paw Pulse Oximeter Sensor
For use with PhysioSuite and PhysioSuite Modules.
$970.90 - $1616.31
Specifications
| Animal | Rat |
|---|---|
| For Use With (Application) | Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring |
| For Use With (Equipment) | PhysioSuite and PhysioSuite Modules |
Description
- MRI compatible sensor includes 3m cable
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring
Specifications
| Rat | |
| Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring | |
| PhysioSuite and PhysioSuite Modules |