missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Acid Phosphatase, Wheat Germ

Supplier:  MP Biomedicals™ 0215387691

 View more versions of this product

Catalog No. ICN15387691


Explore available promotions

Description

Description

  • Activity 20units/mg

Specifications

Specifications

20 U/mg
90%
Catalyzes the hydrolysis of Monoesters and Anhydrides of Phosphoric Acid
Wheat germ
Acid Phosphatase
Lyophilized Powder
Brown
4 to 7
-20°C
1 g
58
Videos
SDS
Documents

Documents

Promotions

Promotions