MP Biomedicals™ AIN-93G Mineral Mix, Powder
Powdered, for rodents
Supplier:  MP Biomedicals™ 0296040002
 View more versions of this product
Catalog No. ICN96040002
Explore available promotions
Description
Description
For growth, pregnancy and lactational phases of rodents
Specifications
Specifications
Animal Diet
Powder
2 kg
15 to 30°C
Calcium Carbonate 35.7%, Monopotassium Phosphate 19.6%, Potassium Citrate Monohydrate 7.078%, Sodium Chloride 7.4%, Potassium Sulfate 4.66%, Ma gnesium Oxide 2.4%, Ferric Citrate 0.606%, Zinc Carbonate 0.165%, Man ganese Carbonate 0.063%, Copper Carbonate 0.03%, Potassium Iodate 0.001%, Sodium Selenate, Anhydrous 0.00103%, Ammonium Molybdate.4H2O 0.000795%, Sodium Metasilicate.9H2O 0.145%, Chromium Potassium Sulfate.12H2O 0.0275%, Lithium Chloride 0.00174%, Boric Acid 0.008145%, Sodium Fluoride 0.00635%, Nickel Carbonate 0.00318%, Ammonium Vanadate 0.00066%, Powdered Su gar 22.1%
Growth, pregnancy and lactational phases of Rodents
Rodent
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.