missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on carboxyl end of the bond
Supplier: MP Biomedicals™ 0215227205
Description
α-Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan.
- CAS Number: 9004-07-3
- EC Number: 232-671-2
- MDL Number: MFCD00130481
- Extinction Coefficient: 20.4
- Format: white powder
- From Bovine Pancreas
- Crystallized, dialyzed, salt-free lyophilized powder
Spécifications
1X | |
Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan | |
Bovine pancreas | |
alpha-Chymotrypsin | |
Heavy Metals, natural Trypsin inhibitors to various degrees, Organophosphorus compounds, AEBSF, alpha-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and a-2-Macroglobulin |
White | |
0°C | |
5 g | |
25 | |
Crystallized, Lyophilized |