missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas

A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on carboxyl end of the bond

Supplier:  MP Biomedicals™ 021522722

 View more versions of this product

Catalog No. ICN1522722


Explore available promotions

Description

Description

α-Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan.

  • CAS Number: 9004-07-3
  • EC Number: 232-671-2
  • MDL Number: MFCD00130481
  • Extinction Coefficient: 20.4
  • Format: white powder
  • From Bovine Pancreas
  • Crystallized, dialyzed, salt-free lyophilized powder

Specifications

Specifications

1X
Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan
Bovine pancreas
alpha-Chymotrypsin
Heavy Metals, natural Trypsin inhibitors to various degrees, Organophosphorus compounds, AEBSF, alpha-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and a-2-Macroglobulin
White
0°C
250 mg
25
Crystallized, Lyophilized
Videos
SDS
Documents

Documents

Promotions

Promotions