missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Ampicillin, Sodium Salt

Serves a variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

Supplier:  MP Biomedicals™ 0219452605

 View more versions of this product

Catalog No. ICN19452605


Explore available promotions

Description

Description

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Applications:

Selection antibiotic effective against most gram negative and gram positive bacteria.

Specifications

Specifications

4°C
Antibiotic
8 to 10
Powder
Non-sterile
5 g
Videos
SDS
Documents

Documents

Promotions

Promotions