missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Arginase from Bovine Liver

Major degradative enzyme for arginine that converts arginine to ornithine

Supplier:  MP Biomedicals™ 0210027703

 View more versions of this product

Catalog No. ICN10027703


Explore available promotions

Description

Description

The enzyme participates in the Krebs-Henseleit urea cycle. It is most highly concentrated in mammalian liver and also present in mammary glands.

  • Synonyms: L-arginine amidinase, L-arginine amidino-hydrolase
  • CAS Number: 9000-96-8
  • EC Number: 232-570-3
  • MDL Number: MFCD00081368
  • Format: Brown powder
  • Specific Activity: ∽20U/mg
  • Extinction Coefficient E1%: 278nm
  • pH: Optimum pH: 10 (Mn2+ activated)
  • Dissolves readily at 1mg/mL in 0.05M Manganese maleate (pH 7.0) to give clear pale brown solution
  • Soluble in distilled water or diluted buffer

Specifications

Specifications

1 mg/mL
10
-20°C
3 kU
115000 to 120000
Powder
Brown
Converts Arginine to Ornithine, deficiency is associated with spasticity and motor dysfunction
Bovine liver
Arginase
Hg2+, Ag2+, Zn2+
Videos
SDS
Documents

Documents

Promotions

Promotions