missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Casein sodium salt
Casein sodium salt White to off-white powder
Supplier: MP Biomedicals™ 0290289605
Description
- Casein protein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
- Casein sodium salt from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein.
- Casein is proposed to be the main protective constituent in milk.
Specifications
Casein Sodium salt | |
5 kg | |
Powder | |
Soluble in water (1% aq soln-Cloudy Solution). |
6.5 to 7.5 | |
15° to 30°C | |
White | |
Sodium Caseinate |