missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Cellulase from Aspergillus niger
Cellulase from Aspergillus niger catalyzes hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan and cellooligosaccharides cellotriose to cellohexaose
$230.00 - $762.18
Specifications
Concentration | >60,000 U/g |
---|---|
Color | Tan |
pH | 4.2 to 5.2 |
For Use With (Application) | Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels |
Content And Storage | 4°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15058325
|
MP Biomedicals™
0215058325 |
25 g |
Each for $230.00
|
|
|||||
ICN15058380
|
MP Biomedicals™
0215058380 |
100 g |
Each for $762.18
|
|
|||||
Description
Cellulase refers to a group of enzymes which acting together hydrolyze cellulose. The b-glucosidases, including cellobiose are primarily active on the smaller molecular weight cellulose hydrolysates.
- CAS Number: 9012-54-8
- EC Number: 232-734-4
- MDL Number: MFCD00081510
- Format: Tan powder
- Specific activity: >60000U/g
- Readily soluble in water
- Foreign activity: Salmonella Negative
- Optimum pH: 4.2 to 5.2
Cellulase is used in molecular biology, cell biology, textile industry, pulp and paper industry and for commercial food processing in coffee
Specifications
>60,000 U/g | |
4.2 to 5.2 | |
4°C | |
Cellulase |
Tan | |
Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels | |
Aspergillis niger | |
Powder |