missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ FastDNA™ SPIN Kit for Soil, 50 mL Tubes

FastDNA SPIN Kit for Soil is used for extracting bacterial, fungal, plant, and animal DNA from soil and other environmental samples. It uses Lysing Matrix E for sample processing on a FastPrep™ instrument.

Supplier:  MP Biomedicals™ 116560600

Catalog No. MP116560600


Explore available promotions

Description

Description

  • Product Overview: The FastDNA Spin Kit for Soil is designed to isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples. Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms. The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications. Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
  • Applications: Cited in over 6000 scientific publications, the FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water. The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors
  • Sample Type: Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast
  • Sample Volume: Approximately 200 μL to 50 mL; up to approximately 25 g
Specifications

Specifications

Room Temperature
Use to isolate bacterial, fungi, plant and animal genomic DNA from soil and environmental samples
FastDNA-24 Kits
Seeds, Spores, Animal Tissues, Feces, Food, Fungi, Plants, Roots
10 each Garnet Lysing Matrix Tubes (50 mL), 60 mL PLS Solution, 15 mL MT Buffer, 350 mL Sodium Phosphate Buffer, 25 mL PPS for Soil, 120 mL Binding Matrix, 120 mL GTC Wash Buffer, 12 mL SEWS-M, 60 mL TES, 200μL BBS Gel Loading Dye, 10 each SPIN Filter Tubes (50 mL)
Approximately 200 μL to 50 mL; Up to approximately 25 g
Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.