missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ FastDNA™ SPIN Kit
FastDNA™ SPIN Kit is used for isolation of DNA from up to 200 mg of almost any sample in less than 30 minutes. This is the updated version of the well-known FASTDNA Kit and can be used with any FastPrep Instrument.
Supplier: MP Biomedicals™ 116540600
Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mL tubes, Microcentrifuge tubes (2.0 mL and 1.5 mL), Rotator or low-speed vortex
Description
- Product Overview: The FastDNA™ SPIN Kit, an updated version of the well-known FastDNA™ Kit, is used with any FastPrep™ Instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
- Features: Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin fi lter method: Rapid and reproducible sample lysis with the FastPrep™-24 or FastPrep™ FP120 Instrument, Lyse and isolate DNA in less than 30 minutes from a variety of sample types, Flexible format eliminates need for separate DNA isolation kits, No hazardous organic reagents required, SPIN Filters are included to streamLine silica handling.
- Applications: The Fast DNA™ SPIN Kit, an updated version of the well known Fast DNA TM Kit, is used with the FastPrep™-24 or FastPrep™ FP120 Instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
- Sample Type: Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Wastewater, Yeast
- Sample Volume: Up to 200 mg per prep
Specifications
| Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants | |
| Lysing Matrix A 100 x 2 mL Tubes, 100 x 0.25 in. Ceramic Spheres, 66 mL Binding Matrix, 12 mL Concentrated SEWS-M, 25 mL DES, 90 mL CLS-VF, 25 mL PPS, 110 mL CLS-TC, 110 mL CLS-Y, 100 x SPIN modules, 100 x Catch Tubes, 1 x User Manual, 1 x MSDS | |
| FastDNA-24 Kits |
| Isolates genomic DNA from plants, animals, bacteria, yeast, algae and fungi | |
| 100 Preps | |
| Up to 200 mg per prep |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.