MP Biomedicals™ Human Albumin
From human plasma
Supplier:  MP Biomedicals™ 0219134905
 View more versions of this product
Catalog No. ICN19134905
Explore available promotions
Description
Description
Negative for HBsAg and HIV antibodies as tested. Purity: >97%. Salt-free, lyophilized, homogeneous powder. MW: 65kDa.
Specifications
Specifications
ELISA, Molecular Biology, Western Blot
Human Albumin
Human
Lyophilized
65
5g
4°C